United Electric E100-8BC Nema 4x Temperature Switch, 350/640F; Remote Mount
Representative image only.
United Electric E100-8BC Nema 4x Temperature Switch, 350/640F; Remote Mount
United Electric | Mfr: E100-8BC| Item: 3900822
₹137,000.00INR / Each
₹137,000.00INR / Each
Product Description
Key Features
  • 2 year warranty
More About this Item
FEATURES
  • Legendary UE reliability
  • Wideselection of temperature ranges
  • Internal reference scales and adjustment lock
  • Enclosure design permits safe and easy access for wiring
  • Choice of local or remote mount
  • NEMA 4X die cast aluminum enclosure, epoxy powder coated, gasketed, captive screws
Two year warranty
Specifications
  • RelaysSPDT
  • Min Temperature (° F)350
  • Max Temperature (° F)640
  • DescriptionNema 4x Temperature Switch, 350/640F; Remote Mount