Unico Series Spectrophotometer Software
Representative image only.
Unico Series Spectrophotometer Software
Unico | Mfr: IS215-401| Item: 8305765
Customize your spectrophotometer
₹75,700.00INR / Each
₹75,700.00INR / Each
Product Description
Key Features
  • Compatible with Unico visible spectrophotometers
  • Easy to install and connect
  • Ensure accuracy and precision
  • 1 year warranty
More About this Item
Customize your Unico spectrophotometer with cell holders, sample chambers, and other accessories.
Standard Curve, Abs / %T / conc., Abs vs. time; for Windows® XP.
Software, Graphics, for 83057-Series Spectrophotometers.
1 Year Warranty
Specifications
  • Compatible With83057-01, 83057-06, 83059-10, 83059-15
  • DescriptionSeries Spectrophotometer Software