Unico Flow Cell for Spectrophotomer Sipper Systems, Quartz
Representative image only.
Unico Flow Cell for Spectrophotomer Sipper Systems, Quartz
Unico | Mfr: S-90-346FQ| Item: 8305779
Customize your spectrophotometer
₹162,000.00INR / Each
₹162,000.00INR / Each
Product Description
Key Features
  • Compatible with Unico visible spectrophotometers
  • Easy to install and connect
  • Ensure accuracy and precision
  • 1 year warranty
More About this Item
Customize your Unico spectrophotometer with cell holders, sample chambers, and other accessories.
Required for Unico Spectrophotometer Sipper Systems, or Combination Temperature Control/Sipper System. Quartz cell is intended for UV light measurements.
1 Year Warranty
Specifications
  • MaterialQuartz
  • Compatible With83059-10/83059-15
  • DescriptionFlow Cell for Spectrophotomer Sipper Systems, Quartz