Ultraviolet Germicidal Fluorescent Replacement Bulb for Eisco Labs Goggle Sanitizing Cabinet, 25 watt, TUV
Representative image only.
Ultraviolet Germicidal Fluorescent Replacement Bulb for Eisco Labs Goggle Sanitizing Cabinet, 25 watt, TUV
| Mfr: GGSN10BULB| Item: 8647807
Ideal replacement bulb for your goggle sanitizer cabinet
₹11,300.00INR / Each
₹11,300.00INR / Each
Product Description
Key Features
  • Produces UVC radiation to kill microscopic organisms
  • Offers up to 8000 working hours
  • EPA Approved
  • 1 year parts & labor warranty
More About this Item
Keep your Eisco Lab goggle sanitizer cabinet running and dependable with this ultraviolet germicidal fluoresecent replacement bulb. The bulb offers constant UV output over its complete lifetime, for maximum security of disinfection and high system efficacy.
1 Year Parts and Labor
Specifications
  • Bulb / Tube TypeT8
  • Bulb / Tube Life8000 hours
  • Power (watts)25
  • DescriptionGermicidal Fluoresecent Replacement Bulb for Eisco Labs Goggle Sanitizing Cabinet, 25 watt, TUV
  • Warranty1 year parts & labor
  • Length (InchShort)18
  • Length (cm)45.72