Tunair SS-3017 Silicone Membrane Cap Liner, for 2.5 L Flasks; 5/Pk

Representative image only.

Tunair SS-3017 Silicone Membrane Cap Liner, for 2.5 L Flasks; 5/Pk

Tunair | Mfr: SS-3017| Item: 0183593

Optimize peformance of your Tunair shaking flask

₹43,200.00INR / Pkg of 5

₹43,200.00INR / Pkg of 5
Product Description

More About this Item

Replacement components will enhance the life of your shaker flask.

Specifications
  • Capacity (mL)2500
  • Cap MaterialSilicone
  • DescriptionSilicone Membrane Cap Liner, for 2.5 L Flasks; 5/Pk
Reviews
Q&A