Tube Insert, PP, 5/16" OD; 10/Pk
Representative image only.
Tube Insert, PP, 5/16" OD; 10/Pk
| Item: 3623494
For supporting soft or flexible tubing in compression fitting assemblies
₹830.00INR / Pkg of 10
₹830.00INR / Pkg of 10
Product Description
Key Features
  • Polypropylene tube inserts reinforce soft or thin-wall plastic tubing
  • Help prevent tube collapse at the compression connection
  • Polypropylene offers good resistance to alkalis, salts, and most weak acids
  • Material temperature rating: -34 to 107°C (-30 to 225°F)
  • NSF 14 certified, NSF 61 compliant, and autoclavable
  • 5/16" OD
More About this Item
JACO polypropylene tube inserts are designed to fit inside soft or flexible plastic tubing, reinforcing the tube wall at the connection point. The insert helps prevent tubing from collapsing under compression and supports a more consistent seal and grip.
These inserts are a practical choice for water-treatment, filtration, DI water, and mild chemical-feed tubing assemblies where soft or thin-wall tubing needs added internal support. Polypropylene is lightweight, rigid, and resistant to environmental stress cracking, with good chemical resistance for applications involving alkalis, salts, and most weak acids.
The inserts have a solid opaque, flat white appearance, a material temperature rating of -34 to 107°C (-30 to 225°F), and are NSF 14 certified, NSF 61 compliant, and autoclavable.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPP
  • Compression Size / Tube Od (Port 1) (in)5/16
  • Compression Size / Tube Od (Port 2) (in)5/16
  • ColorNatural
  • Min Temperature (° F)-30
  • Max Temperature (° F)225
  • Min Temperature (° C)-34
  • Max Temperature (° C)107
  • Quantity10
  • DescriptionTube Insert, PP, 5/16" OD; 10/Pk