TSI Alnor 801750 Air Cone Flow Hood Accessory Kit
Representative image only.
TSI Alnor 801750 Air Cone Flow Hood Accessory Kit
TSI | Mfr: 801750| Item: 1037449
Optimize performance of your air velocity meters
₹85,200.00INR / Each
₹85,200.00INR / Each
Product Description
Key Features
  • Kit comes with both rectangular and circular cones
  • Measures volumetric flow at grilles, diffusers and registers
  • Reads air volume quickly and accurately
  • Excellent choice for small grilles
  • Works with Airflow LCA301 and LCA501 rotating vanes
More About this Item
Parts and accessories to meet your application needs.
Aircone Flow Hoods are a fast and accurate method of maximizing the usefulness of your 4" (100 mm) rotating vane anemometers. You can double the capability of your rotating vane, turning it into an air volume flow balancing tool.
The range (with LCA301) is 0 to 0.1 m3/s (0 to 210 cfm). The rectangular hood size is 285 mm x 235 mm (11.2" x 9.2") while the round hood size is 180 mm (7.1") diameter.
Specifications
  • Max Air Volume (CFM)210
  • Min Air Volume (CFM)0
  • DescriptionAir Cone Flow Hood Accessory Kit