TSI Alnor 801206 Hood Expansion Kit
Representative image only.
TSI Alnor 801206 Hood Expansion Kit
TSI | Mfr: 801206| Item: 1037497
Optimize performance of your air velocity meters
₹180,000.00INR / Each
₹180,000.00INR / Each
Product Description
Key Features
  • Extend the size of your barometer for large surface area measurment
  • 1 year warranty
More About this Item
Parts and accessories to meet your application needs.
This hood expansion kit allows your to extend the size of your balometer. This kit comes with 1' X 4' (305mm X 1220mm) and 2' X 4' (610mm X 1220mm) hood and air frames. This extension kit is compatible with TSI models EBT731 and ABT701.
1 Year Warranty
Specifications
  • DescriptionHood Expansion Kit (1' X 4' (305mm X 1220mm) and 2' X 4' (610mm X 1220mm) hoods)