TSI Alnor 634634004 Telescoping Pitot Tube
Representative image only.
TSI Alnor 634634004 Telescoping Pitot Tube
TSI | Mfr: 634634004| Item: 1030245
Optimize performance of your air velocity meters
₹102,000.00INR / Each
₹102,000.00INR / Each
Product Description
Key Features
  • Obtain multiple lengths with one pitot tube
  • No more changing out pitot tubes for various lenghts!
More About this Item
Parts and accessories to meet your application needs.
This telescoping pitot probe has a length range of 8" to 38". It is best suited for HVAC commissioning where multiple pitot lengths are required, or as a means of replacing several pitot tubes with a single product. For use with TSI DP-Calc micromanometers.
Specifications
  • Probe length (in)38
  • DescriptionTelescoping Pitot Tube