TSI 801090 Velocity Matrix, 16 point with Telescopic Handle and Tubing
Representative image only.
TSI 801090 Velocity Matrix, 16 point with Telescopic Handle and Tubing
TSI | Mfr: 801090| Item: 0594942
Optimize performance of your air velocity meters
₹202,000.00INR / Each
₹202,000.00INR / Each
Product Description
Includes
velocity stand-offs, telescopic handle, and two 8' hoses.
Key Features
  • Averages 16-point air volume/velocity measurement
  • Air velocity range of 25 to 2500 ft/min (0.125 to 12.5 m/s)
  • Ideal for measuring the face velocities of fume hoods, filters, and other large surface area applications
  • 2 year warranty
More About this Item
Parts and accessories to meet your application needs.
The grid covers 1 cubic foot (0.09 m2) and averages the air velocity while minimizing the effects of turbulence to produce a stale reading. This velocity probe is compatible with TSI-Alnor Capture Hood Models EBT730 and EBT731; TSI-Airflow Capture Hood Models PH730 and PH731; and TSI Capture Hood Models 8380 and 8715.
Two year warranty
Specifications
  • Max Air Velocity (ft/min)2500
  • Min Air Velocity (ft/min)25
  • Width (in)12
  • Length (in)12
  • DescriptionVelocity Matrix, 16 point with Telescopic Handle and Tubing