Troemner WA061 Calibration Weight Handling Cotton Gloves, 1 Each
Representative image only.
Troemner WA061 Calibration Weight Handling Cotton Gloves, 1 Each
Troemner | Mfr: WA061| Item: 0102818
₹2,300.00INR / Each
₹2,300.00INR / Each
Product Description
More About this Item
Leather gloves are designed for large industrial weights and are sold as a single glove.
Nylon gloves are sold as a pair and feature a grip tip for secure lifting.
Cotton gloves are sold individually and are designed to fit either the left or right hand.
Specifications
  • MaterialCotton
  • DescriptionCalibration Weight Handling Cotton Gloves, 1 Each