Troemner WA056 Polycarbonate Tweezer for Weights 1mg, Through 200G
Representative image only.
Troemner WA056 Polycarbonate Tweezer for Weights 1mg, Through 200G
Troemner | Mfr: WA056| Item: 0102788
₹5,400.00INR / Each
₹5,400.00INR / Each
Product Description
More About this Item
Forceps / Tweezers are designed for proper handling of weights.
Stainless steel forceps are available in 5” and 8” sizes and are designed in a smooth tip, curved tip, straight grip tip, and a 22° grip tip. Stainless steel forceps tips are made of PVC.
Economical polycarbonate forceps for weights 1 mg through 200 g.
Specifications
  • Overall Length (in)4
  • Overall Length (cm)10.2
  • MaterialPolycarbonate
  • DescriptionTroemner WA056 Polycarbonate Tweezer for Weights 1mg, Through 200G