Troemner TW-5T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 lb to 1/32 oz
Representative image only.
Troemner TW-5T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 lb to 1/32 oz
Troemner | Mfr: TW-5T| Item: 0910899
Stainless Steel Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians.
₹300,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹300,000.00INR / Each
Buy It Together
Current Item
Troemner TW-5T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 lb to 1/32 oz
Troemner TW-5T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 lb to 1/32 oz
0910899
₹300,000.00INR / Each
+ + =
Total Price 413300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
all weights include noted documentation of accuracy from the vendor and case
Key Features
  • 1 year warranty
More About this Item
"Small' Weight sets where the total weight is less than 15 lb are supplied in sturdy plastic cases with a carrying handle.
"Large" Weight sets where the total weight is greater than 15 lb are supplied in a case with metal reinforced corners and bottoms with padded handles for easier carrying.
Weights 8 oz and above each have their own compartment within the dense foam interior. Smaller weights are contained within a smaller synthetic leather case that fits into the larger “master” carrying case. This “case-in-a-case” design helps keep smaller weights from getting lost and also allows smaller weights to be taken out of the master case and used closer to the user’s application.
1 Year Warranty
Specifications
  • Mass ClassClass F
  • Mass Size (lbs)5
  • Mass CertificationNIST/NVLAP
  • Contents of Set5lb, (2) 2lb, 1lb, 8oz, 4oz, 2oz, 1oz, 1/2oz, 1/4oz, 1/8oz, 1/16oz, (2) 1/32oz
  • Description5 Lb-1/32 oz Class F Stainless Steel Test Weight Set with NIST Certificate