Troemner TW-5000-01T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 kg to 1 mg
Representative image only.
Troemner TW-5000-01T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 kg to 1 mg
Troemner | Mfr: TW-5000-01T| Item: 0910895
Stainless Steel Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians.
₹306,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹306,000.00INR / Each
Buy It Together
Current Item
Troemner TW-5000-01T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 kg to 1 mg
Troemner TW-5000-01T Class F Stainless Steel Test Weight Set with NIST Calibration Certificate; 5 kg to 1 mg
0910895
₹306,000.00INR / Each
+ + =
Total Price 391300.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
all weights include noted documentation of accuracy from the vendor and case
Key Features
  • 1 year warranty
More About this Item
"Small' Weight sets where the total weight is less than 15 lb are supplied in sturdy plastic cases with a carrying handle.
"Large" Weight sets where the total weight is greater than 15 lb are supplied in a case with metal reinforced corners and bottoms with padded handles for easier carrying.
Weights 8 oz and above each have their own compartment within the dense foam interior. Smaller weights are contained within a smaller synthetic leather case that fits into the larger “master” carrying case. This “case-in-a-case” design helps keep smaller weights from getting lost and also allows smaller weights to be taken out of the master case and used closer to the user’s application.
1 Year Warranty
Specifications
  • Mass ClassClass F
  • Mass Size (kg)5
  • Mass CertificationNIST/NVLAP
  • Contents of Set5000g, (2) 2000g, 1000g, 500g, (2) 200g, 100g, 50g, (2) 20g, 10g, 5g, (2) 2g, 1g, 500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg, 5mg, (2) 2mg, 1mg
  • Description5kg.-1mg. Class F Stainless Steel Test Weight Set with NIST Certificate,