Troemner 9484T 5 lb, 36" Avoirdupois Weight Hanger with NIST Traceable Certifiate
Representative image only.
Troemner 9484T 5 lb, 36" Avoirdupois Weight Hanger with NIST Traceable Certifiate
Troemner | Mfr: 9484T| Item: 1196683
Use in a variety of applications such as pressure, torque, and tensile strength testing
₹88,200.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹88,200.00INR / Each
Buy It Together
Current Item
Troemner 9484T 5 lb, 36" Avoirdupois Weight Hanger with NIST Traceable Certifiate
Troemner 9484T 5 lb, 36" Avoirdupois Weight Hanger with NIST Traceable Certifiate
1196683
₹88,200.00INR / Each
+ =
Total Price 105700.00 ADD 2 ITEMS TO CART
Product Description
Includes
noted documentation of accuracy from the vendor, not supplied in a case
Key Features
  • Color coded – silver for avoirdupois
  • 3 month warranty
More About this Item
Cast Iron Slotted Weights are typically used with a hanger that also has its weight calibrated so the hanger can be used as part of the overall weight under test. Weight hangers are available in a variety of lengths and weight capacities. Hangers are calibrated to a mass value, and also
have a capacity of how much weight can be loaded onto them.
To determine which hanger will accommodate your weights, use the stem height of the hanger
as the maximum height, then use the thickness dimension of each weight along with the quantity you intend to place on the hanger to determine the total height of your stack of weights.
The height of your stack of weights should be less than the length of the hanger. The dimension of the hook opening is ½ inch.
3 Month Warranty
Specifications
  • Mass ClassClass 6
  • Mass Size (lbs)5
  • Mass CertificationNIST
  • Height (in)36
  • Description5 lb, Avoirdupois Weight Hanger with NIST Traceable Certificate