Troemner 313S Economical S.S. Class 7 Weight Set; 1000 g to 1 g

Representative image only.

Troemner 313S Economical S.S. Class 7 Weight Set; 1000 g to 1 g

Troemner | Mfr: 313S| Item: 0103500

Make the most of your balance—calibrate regularly!

₹49,400.00INR / Set of 1

₹49,400.00INR / Set of 1

Buy It Together

Current Item

Troemner 313S Economical S.S. Class 7 Weight Set; 1000 g to 1 g
Troemner 313S Economical S.S. Class 7 Weight Set; 1000 g to 1 g
0103500

₹49,400.00INR / Set of 1
+ + +
Reagents Hydrochloric Acid, 50% (v/v), 500mL, Volumetric
7891158

₹10,800.00INR / Each
+
Reagents Potassium Iodide, 25% (w/v), 1L, Volumetric
7891805

₹24,900.00INR / Each
+
LabChem Nitric Acid, ACS Grade; 1 L
8002952

Call For Price
+
LabChem Potassium Iodide, 10% (w/v); 500 mL
8003513

Call For Price
+
LabChem Sodium Hydroxide, 10% (w/w); 1 L
8003839

Call For Price
+ + =
Total Price 317000.00 ADD 10 ITEMS TO CART
Product Description

Includes

Hinged polypropylene box with snap-tight closure.

Key Features

  • Adjusted to ANSI/ASTM E617 Class 7
  • 7-port manifold assembly
  • Max pressure 1000 psi (69 bar)
  • 0.040" thru-hole diameter
  • 42 µL swept volume
  • 1 year warranty

More About this Item

This economical set is ideal for general-purpose applications. Weight sets are designed to achieve any total mass by combining smaller masses. All weighs feaature a satin finish surface and are marked with their nominal mass value.

Hinged polypropylene box with snap-tight closure contains the following masses (tolerances): 1 g (±5 mg), two 2 g (±5 mg), 5 g (±10 mg), 10 g (±20 mg), two 20 g (±20 mg), 50 g (±30 mg), 100 g (±30 mg), two 200 g (±50 mg), 500 g (±100 mg), and 1000 g (±200 mg). Total of 13 masses.

1 Year Warranty
Specifications
  • Mass ClassClass 7
  • Mass CertificationNone
  • Height (in)1.5
  • Width (in)3.8437
  • Length (in)7.7187
  • Contents of Set1000g, 500g, (2) 200g, 100g, 50g, (2) 20g, 10g, 5g, (2) 2g, 1g; polycarbonate case
  • DescriptionEconomical S.S. Class 7 Weight Set; 1000 g to 1 g
  • Warranty1 year
Reviews
Q&A