Troemner 312-10S Economical S.S. Class 7 Weight Set; 500 g to 10 mg

Representative image only.

Troemner 312-10S Economical S.S. Class 7 Weight Set; 500 g to 10 mg

Troemner | Mfr: 312-10S| Item: 0102897

Make the most of your balance—calibrate regularly!

₹33,300.00INR / Each

₹33,300.00INR / Each

Buy It Together

Current Item

Troemner 312-10S Economical S.S. Class 7 Weight Set; 500 g to 10 mg
Troemner 312-10S Economical S.S. Class 7 Weight Set; 500 g to 10 mg
0102897

₹33,300.00INR / Each
+ + +
Reagents Hydrochloric Acid, 50% (v/v), 500mL, Volumetric
7891158

₹10,800.00INR / Each
+
Reagents Potassium Iodide, 25% (w/v), 1L, Volumetric
7891805

₹24,900.00INR / Each
+
LabChem Nitric Acid, ACS Grade; 1 L
8002952

Call For Price
+
LabChem Potassium Iodide, 10% (w/v); 500 mL
8003513

Call For Price
+
LabChem Sodium Hydroxide, 10% (w/w); 1 L
8003839

Call For Price
+ + =
Total Price 300900.00 ADD 10 ITEMS TO CART
Product Description

Includes

Hinged polypropylene box with snap-tight closure.

Key Features

  • Adjusted to ANSI/ASTM E617 Class 7

More About this Item

This economical set is ideal for general-purpose applications. Weight sets are designed to achieve any total mass by combining smaller masses. All weighs feaature a satin finish surface and are marked with their nominal mass value.

Specifications
  • Mass ClassClass 7
  • Mass CertificationNone
  • Height (in)1.5
  • Width (in)3.8437
  • Length (in)7.7187
  • Contents of Set500g, (2) 200g, 100g, 50g, (2) 20g, 10g, 5g, (2) 2g, 1g, 500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg; polycarbonate case
  • DescriptionEconomical S.S. Class 7 Weight Set; 500 g to 10 mg
Reviews
Q&A