Troemner 300S Economical S.S. Class 7 Weight Set; 500 g to 1 g

Representative image only.

Troemner 300S Economical S.S. Class 7 Weight Set; 500 g to 1 g

Troemner | Mfr: 300S| Item: 0102887

Make the most of your balance—calibrate regularly!

₹29,000.00INR / Each

₹29,000.00INR / Each

Buy It Together

Current Item

Troemner 300S Economical S.S. Class 7 Weight Set; 500 g to 1 g
Troemner 300S Economical S.S. Class 7 Weight Set; 500 g to 1 g
0102887

₹29,000.00INR / Each
+ + +
Reagents Hydrochloric Acid, 50% (v/v), 500mL, Volumetric
7891158

₹10,800.00INR / Each
+
Reagents Potassium Iodide, 25% (w/v), 1L, Volumetric
7891805

₹24,900.00INR / Each
+
LabChem Nitric Acid, ACS Grade; 1 L
8002952

Call For Price
+
LabChem Potassium Iodide, 10% (w/v); 500 mL
8003513

Call For Price
+
LabChem Sodium Hydroxide, 10% (w/w); 1 L
8003839

Call For Price
+ + =
Total Price 296600.00 ADD 10 ITEMS TO CART
Product Description

Key Features

  • Adjusted to ANSI/ASTM E617 Class 7

More About this Item

This economical set is ideal for general-purpose applications. Weight sets are designed to achieve any total mass by combining smaller masses. All weighs feaature a satin finish surface and are marked with their nominal mass value.

Specifications
  • Mass ClassClass 7
  • Mass CertificationNone
  • Height (in)1.7812
  • Width (in)2.375
  • Length (in)5.625
  • Contents of Set500g, 200g, (2) 100g, 50g, 20g, (2) 10g, 5g, (2) 2g, 1g
  • DescriptionEconomical S.S. Class 7 Weight Set; 500 g to 1 g
Reviews
Q&A