Troemner 1372T 10kg Class-F Stainless Steel Test Weight, Traceable Certificate, Grip Handle
Representative image only.
Troemner 1372T 10kg Class-F Stainless Steel Test Weight, Traceable Certificate, Grip Handle
Troemner | Mfr: 1372T| Item: 1196651
Stainless Steel Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians.
₹183,000.00INR / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
₹183,000.00INR / Each
Buy It Together
Current Item
Troemner 1372T 10kg Class-F Stainless Steel Test Weight, Traceable Certificate, Grip Handle
Troemner 1372T 10kg Class-F Stainless Steel Test Weight, Traceable Certificate, Grip Handle
1196651
₹183,000.00INR / Each
+ =
Total Price 200500.00 ADD 2 ITEMS TO CART
Product Description
Includes
all weights include noted documentation of accuracy from the vendor, not supplied in a case
Key Features
  • 3 month warranty
More About this Item
Stainless Steel Grip Handle Weights are legal for trade, which means their design meets specifications as outlined in NIST Handbook 105-1 and are approved to be used on legal for trade devices within the United States. Weights 5 kg/10 lb and below must be made of stainless steel of quality no lower than 300 series. Adjusting cavities are typically on the top of the weight and must be tamper resistant.
Stainless Steel Class F Test Weights may be used to test most accuracy Class III scales, all scales of Class IIIL or IIII, and scales not marked with a class designation. Class F is also acceptable
for use in verifying scales that have 0.1% accuracy or lower. Class F Test Weights are primarily used to test commercial weighing devices by state and local weights and measures officials, device installers, and service technicians. Suggested markets for Stainless Steel Test Weights include, but are not limited to, Hospitals and Food Services.
3 Month Warranty
Specifications
  • Mass ClassClass F
  • Mass Size (kg)10
  • Mass Size (lbs)22
  • Mass CertificationNIST
  • Description10kg Class F Stainless Steel Test Weight, Traceable Certificate, Grip Handle