TrippNT MRI Locking Lab Storage Cart, Narrow, White
Representative image only.
TrippNT MRI Locking Lab Storage Cart, Narrow, White
TrippNT | Mfr: 50729| Item: 6672013
Secure, flexible storage that is MR safe
₹370,000.00INR / Each
₹370,000.00INR / Each
Buy It Together
Current Item
TrippNT MRI Locking Lab Storage Cart, Narrow, White
TrippNT MRI Locking Lab Storage Cart, Narrow, White
6672013
₹370,000.00INR / Each
+ =
Total Price 445900.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Promotes safety by reducing clutter
  • Constructed of MRI safe materials
  • Flexible storage for bottles, gloves, wipers, and more
  • Keep MRI supplies mobile and organized
  • Non-ferrous HDPE and ABS construction is MRI safe
  • Roll top tambour door provides a space saving design
  • Limited Lifetime Warranty
More About this Item
This TrippNT lab storage furniture is compatible for use within the sensitive magnetic imaging environment, keeping supplies mobile and organized. Featuring white, non-ferrous, durable construction, these lab organizers reduce clutter with a sleek design. Choose from a wide range of shelves and carts that is sure to fit your needs in the lab.
Limited Lifetime Warranty
Specifications
  • Width (in)12
  • Height (in)35
  • Depth (in)19
  • Number of Shelves0
  • Number of Bins0
  • MaterialMRI suitable HDPE and ABS
  • ColorWhite
  • Number of Drawers6
  • DescriptionMRI Locking Lab Storage Cart, Narrow, White
  • WarrantyLimited Lifetime