TrippNT MRI Lab Storage Shelf with 5 Bins, White
Representative image only.
TrippNT MRI Lab Storage Shelf with 5 Bins, White
TrippNT | Mfr: 51888| Item: 6672008
Secure, flexible storage that is MR safe
₹45,300.00INR / Each
₹45,300.00INR / Each
Buy It Together
Current Item
TrippNT MRI Lab Storage Shelf with 5 Bins, White
TrippNT MRI Lab Storage Shelf with 5 Bins, White
6672008
₹45,300.00INR / Each
+ =
Total Price 121200.00 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Promotes safety by reducing clutter
  • Constructed of MRI safe materials
  • Flexible storage for bottles, gloves, wipers, and more
  • Three shelves with five storage areas safety store bottles, gloves, wipers, or chemicals
  • PVC construction is MRI safe
  • Limited Lifetime Warranty
More About this Item
This TrippNT lab storage furniture is compatible for use within the sensitive magnetic imaging environment, keeping supplies mobile and organized. Featuring white, non-ferrous, durable construction, these lab organizers reduce clutter with a sleek design. Choose from a wide range of shelves and carts that is sure to fit your needs in the lab.
Limited Lifetime Warranty
Specifications
  • Width (in)12
  • Height (in)16
  • Depth (in)10
  • Number of Shelves3
  • Number of Bins5
  • MaterialMRI suitable PVC
  • ColorWhite
  • Number of Drawers0
  • DescriptionMRI Lab Storage Shelf with 5 Bins, White
  • WarrantyLimited Lifetime