TriForest BPC0125 Square Autoclavable Media Bottle, 125 mL, PC, 24 per pack, 144/CS

Representative image only.

TriForest BPC0125 Square Autoclavable Media Bottle, 125 mL, PC, 24 per pack, 144/CS

TriForest | Mfr: BPC0125| Item: 0654221

Withstand multiple autoclave cycles

₹112,000.00INR / Case of 144

₹112,000.00INR / Case of 144
Product Description

Includes

leakproof caps (installed).

Key Features

  • Polycarbonate (PC) is a safe and shatterproof alternative to glass
  • Manufactured in an ISO Class 7 (Class 10,000) cleanroom in an ISO 9001 compliant facility
  • Keep samples secure—leakproof seals included
  • Easily complete fill with molded-in graduations
  • 24 bottles per pack, 6 packs per case; 144 bottles total

More About this Item

Store buffers, reagents, media, and other critical fluids in these sturdy, clear bottles. The square shape saves shelf space by allowing bottles to be placed closer together.

Bottles come separated into packs by bags, and a case is double bagged to maintain cleanliness.

Autoclavable

ISO9001 Certified Supplier
Specifications
  • Capacity (mL)125
  • Capacity (oz)4
  • MaterialPolycarbonate(PC)
  • Min Graduation (mL)25
  • Max Graduation (mL)125
  • Graduation Interval (mL)25
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size38/430
  • Cap ColorBlue
  • Bottle ColorTransparent
  • Special FeaturesPacked in bags
  • AutoclavableYes
  • DescriptionSquare/Octagonal Media Bottle, 125 mL; 144/Cs
Reviews
Q&A