Trajan SGE 215145 Vespel®-graphite 85/15 Ferrule with 2 Holes, for 1/16" Fitting, 0.5 x 0.4 mm Hole, 3.2 mm W, 3.7 mm L; 10/Pk
Representative image only.
Trajan SGE 215145 Vespel®-graphite 85/15 Ferrule with 2 Holes, for 1/16" Fitting, 0.5 x 0.4 mm Hole, 3.2 mm W, 3.7 mm L; 10/Pk
Trajan | Mfr: 215145| Item: 1600776
Graphitised Vespel ferrules for GC/MS
₹10,300.00INR / Pkg of 10
₹10,300.00INR / Pkg of 10
Product Description
Key Features
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE GVF Ferrule with 2 Holes, for 1/16" Fitting, 0.5 x 0.4 mm Hole, 3.2 mm W, 3.7 mm L; 10/Pk