Trajan SGE 213007 ProteCol™ PEEK Ferrules, 1/16" to 1/32"; 5/Pk
Representative image only.
Trajan SGE 213007 ProteCol™ PEEK Ferrules, 1/16" to 1/32"; 5/Pk
Trajan | Mfr: 213007| Item: 1599766
Perfect connections, every time
₹6,200.00INR / Pkg of 5
₹6,200.00INR / Pkg of 5
Product Description
Key Features
  • Zero dead colume for no peak dispersion
  • Available in stainless steel or PEEK
  • Reusable PEEK ferrules, with replacements available
  • 6 month warranty
More About this Item
Allows for connection of any combination of 1/32" PEEKsil tubing, 1/16" PEEKsil tubing, 0.36mm fused silica tubing
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE ProteCol™ PEEK Ferrules, 1/16" to 1/32"; 5/Pk
  • Warranty6 month warranty