Trajan SGE 206102 EasyLOK Fitting with 2 fittings, 2 Ferrules; 2/Pk
Representative image only.
Trajan SGE 206102 EasyLOK Fitting with 2 fittings, 2 Ferrules; 2/Pk
Trajan | Mfr: 206102| Item: 1599756
Quick and Easy HPLC connections
₹5,800.00INR / Pkg of 2
₹5,800.00INR / Pkg of 2
Product Description
Key Features
  • Double ended ferrule seals at two points
  • Knurled to make finger tightening easy
  • 6 month warranty
More About this Item
EasyLok™ fittings are composed of a knurled stainless steel nut and a double ended PEEK ferrule. The PEEK ferrule simply slides over any 1/16” OD tubing to its required position, while the nut is finger tightened. Unlike stainless steel, the PEEK ferrule will not crush the tubing and can be easily readjusted for quick column changes.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE EasyLOK Fitting with 2 fittings, 2 Ferrules; 2/Pk