Trajan SGE 1236104 On/Off Valve with Pneumatic Operation, T Flow Path with Dual Outlet, 100 mm Stem; 1/Ea
Representative image only.
Trajan SGE 1236104 On/Off Valve with Pneumatic Operation, T Flow Path with Dual Outlet, 100 mm Stem; 1/Ea
Trajan | Mfr: 1236104| Item: 1599820
Offered in micro control or pneumatic: delivering gas control in the most appropriate format for your analysis
₹61,300.00INR / Each
₹61,300.00INR / Each
Product Description
Key Features
  • Minimal exposure to stainless steel
  • Fully swept flow path
  • Easy installation
  • Minimal dead volume
  • 6 month warranty
More About this Item
Designed specifically for chromatography applications. Can be used at elevated temperatures from high vacuum to medium pressure.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE On/Off Valve with Pneumatic Operation, T Flow Path with Dual Outlet, 100 mm Stem; 1/Ea