Trajan SGE 103431 Mini Union, Stainless Steel, Glass Lined, 0.4 mm ID; 1/Ea
Representative image only.
Trajan SGE 103431 Mini Union, Stainless Steel, Glass Lined, 0.4 mm ID; 1/Ea
Trajan | Mfr: 103431| Item: 1599717
Available in two material types: stainless steel and Glass Lined Tubing
₹7,700.00INR / Each
₹7,700.00INR / Each
Product Description
Includes
Supplied with the appropriate nuts, ferrules and/or sealing rings
Key Features
  • Stainless steel unions come in a variety of tube sizes and configurations
  • GL T unions are inert and mainly used for connecting fused silica tubing of various internal diameters
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Mini Union, Stainless Steel, Glass Lined, 0.4 mm ID; 1/Ea
  • Warranty6 month warranty