Trajan SGE 103418 Union Tee with Nuts and Ferrules, Stainless Steel, 1/16"x1/16"x1/16"; 1/Ea
Representative image only.
Trajan SGE 103418 Union Tee with Nuts and Ferrules, Stainless Steel, 1/16"x1/16"x1/16"; 1/Ea
Trajan | Mfr: 103418| Item: 1599714
Available in two material types: stainless steel and Glass Lined Tubing
₹8,800.00INR / Each
₹8,800.00INR / Each
Product Description
Includes
Supplied with the appropriate nuts, ferrules and/or sealing rings
Key Features
  • Stainless steel unions come in a variety of tube sizes and configurations
  • GL T unions are inert and mainly used for connecting fused silica tubing of various internal diameters
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Union Tee with Nuts and Ferrules, Stainless Steel, 1/16"x1/16"x1/16"; 1/Ea
  • Warranty6 month warranty