Trajan SGE 092068 Tapered Focus Liner, Compatible with Shimadzu 2010/17A, 3.4 mm ID, 95 mm L; 5/Pk

Representative image only.

Trajan SGE 092068 Tapered Focus Liner, Compatible with Shimadzu 2010/17A, 3.4 mm ID, 95 mm L; 5/Pk

Trajan | Mfr: 092068| Item: 1601002

Spot the Difference in the SGE Lineup!

₹23,900.00INR / Pkg of 5

₹23,900.00INR / Pkg of 5
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Bottom taper minimizes contact with metal parts of the inlet and focuses sample onto the column head. Top taper aids in minimizing sample flashback

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Tapered Focus Liner, Compatible with Shimadzu 2010/17A, 3.4 mm ID, 95 mm L; 5/Pk
  • Warranty6 month warranty
Reviews
Q&A