Trajan SGE 092031 Straight Liners, Compatible with Varian 1078/1079, 0.5 mm ID, 54 mm L; 5/Pk

Representative image only.

Trajan SGE 092031 Straight Liners, Compatible with Varian 1078/1079, 0.5 mm ID, 54 mm L; 5/Pk

Trajan | Mfr: 092031| Item: 1600960

Spot the Difference in the SGE Lineup!

₹22,300.00INR / Pkg of 5

₹22,300.00INR / Pkg of 5
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Straight liners facilitate higher split flows

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Straight Liners, Compatible with Varian 1078/1079, 0.5 mm ID, 54 mm L; 5/Pk
  • Warranty6 month warranty
Reviews
Q&A