Trajan SGE 09200101 Split Straight Liner with Quartz Wool, Compatible with Agilent Systems, 78.5 mm L; 1/Ea

Representative image only.

Trajan SGE 09200101 Split Straight Liner with Quartz Wool, Compatible with Agilent Systems, 78.5 mm L; 1/Ea

Trajan | Mfr: 09200101| Item: 1600921

Spot the Difference in the SGE Lineup!

₹3,200.00INR / Pkg of 1

₹3,200.00INR / Pkg of 1
Product Description

Key Features

  • Color coded by geometry to simplify selection
  • Contains o-rings so you're ready to go
  • Certified deactivation
  • 6 month warranty

More About this Item

Straight liners facilitate higher split flows

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Split Straight Liner with Quartz Wool, Compatible with Agilent Systems, 78.5 mm L; 1/Ea
  • Warranty6 month warranty
Reviews
Q&A