Trajan SGE 090112 Gas and vapor Sampler Replacement Sample Bags; 10/Pk
Representative image only.
Trajan SGE 090112 Gas and vapor Sampler Replacement Sample Bags; 10/Pk
Trajan | Mfr: 090112| Item: 1600919
Ideal for remote sampling of environmental or low pressure stream gases and vapor
₹50,600.00INR / Pkg of 10
₹50,600.00INR / Pkg of 10
Product Description
Includes
Canister, Bulb, 10 bags, marker pen, and instructions.
Key Features
  • Easy to use
  • Ideal for field environmental analysis
  • Resealable sample bags
  • 200 cm3 sample bag capacity
More About this Item
Sampling bags consist of a polyethylene - aluminum foil - polyethylene laminate. The bags are suitable for gas pressure differentials up to 5psi
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • DescriptionGas and vapor Sampler Replacement Sample Bags; 10/Pk