Trajan SGE 090111 Gas and Vapor Sampling Kit with Canister, Bulb, 10 Bags, Marker Pen and Instructions; 1/Ea
Representative image only.
Trajan SGE 090111 Gas and Vapor Sampling Kit with Canister, Bulb, 10 Bags, Marker Pen and Instructions; 1/Ea
Trajan | Mfr: 090111| Item: 1600918
Ideal for remote sampling of environmental or low pressure stream gases and vapor
₹79,400.00INR / Each
₹79,400.00INR / Each
Product Description
Includes
Canister, Bulb, 10 bags, marker pen, and instructions.
Key Features
  • Easy to use
  • Ideal for field environmental analysis
  • Resealable sample bags
  • 200 cm3 sample bag capacity
  • 6 month warranty
More About this Item
Sampling bags consist of a polyethylene - aluminum foil - polyethylene laminate. The bags are suitable for gas pressure differentials up to 5psi
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Gas and Vapor Sampling Kit with Canister, Bulb, 10 Bags, Marker Pen and Instructions; 1/Ea