Trajan SGE 073351 SilTite™ Ferrule Starter Kit, Compatible with 0.32 mm ID Columns; 1/Ea
Representative image only.
Trajan SGE 073351 SilTite™ Ferrule Starter Kit, Compatible with 0.32 mm ID Columns; 1/Ea
Trajan | Mfr: 073351| Item: 1600839
Form a strong and permanent airtight seal aroung the capillary column
₹14,700.00INR / Each
₹14,700.00INR / Each
Product Description
Key Features
  • Strong seal on capillary columns
  • No contamination
  • Leak free
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE SilTite® Ferrule Starter Kit, Compatible with 0.32 mm ID Columns; 1/Ea