Trajan SGE 073228 Shimadzu SilTite™ Ferrules, for 0.32 mm ID Columns; 10/Pk
Representative image only.
Trajan SGE 073228 Shimadzu SilTite™ Ferrules, for 0.32 mm ID Columns; 10/Pk
Trajan | Mfr: 073228| Item: 1600821
Form a strong and permanent airtight seal aroung the capillary column
₹13,800.00INR / Pkg of 10
₹13,800.00INR / Pkg of 10
Product Description
Key Features
  • Strong seal on capillary columns
  • No contamination
  • Leak free
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Shimadzu SilTite® Ferrules, for 0.32 mm ID Columns; 10/Pk