Trajan SGE 073219 SilTite™ Ferrules, Compatible with PerkinElmer 1/32" Columns; 10/Pk
Representative image only.
Trajan SGE 073219 SilTite™ Ferrules, Compatible with PerkinElmer 1/32" Columns; 10/Pk
Trajan | Mfr: 073219| Item: 1600812
Form a strong and permanent airtight seal aroung the capillary column
₹8,600.00INR / Pkg of 10
₹8,600.00INR / Pkg of 10
Product Description
Key Features
  • Strong seal on capillary columns
  • No contamination
  • Leak free
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE SilTite® Ferrules, Compatible with PerkinElmer 1/32" Columns; 10/Pk