Trajan SGE 062803 Capillary Electrophoresis Tubing, Untreated, 363 um OD, 50 um ID, 1 m Length; 1/Ea
Representative image only.
Trajan SGE 062803 Capillary Electrophoresis Tubing, Untreated, 363 um OD, 50 um ID, 1 m Length; 1/Ea
Trajan | Mfr: 062803| Item: 1600693
Precision tubing for performance
₹7,200.00INR / Each
₹7,200.00INR / Each
Product Description
Key Features
  • Manufactured from precision bore tubing
  • hydrophobic and hydrophilic treatments enabling use with proteins, peptides, enzymes
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Capillary Electrophoresis Tubing, Untreated, 363 um OD, 50 um ID, 1 m Length; 1/Ea