Trajan SGE 062712 Fused Silica Capillary Tubing, Non-Deactivated, 363 um OD, 75 um ID, Custom
Representative image only.
Trajan SGE 062712 Fused Silica Capillary Tubing, Non-Deactivated, 363 um OD, 75 um ID, Custom
Trajan | Mfr: 062712| Item: 1600689
Precision Tubing for Performance
₹4,000.00INR / Each
₹4,000.00INR / Each
Product Description
Key Features
  • High homogeneity
  • Chemical inertness
  • High intrinsic tensile strength
  • Pressure resistant to 1000 bar
  • 6 month warranty
More About this Item
Coated fused silica tubing exhibits excellent flexibility, chemical stability, tensile strength and exacting tolerances making it the tubing of choice for GC capillary applications, LC tubing and other micro tubing requirements.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Fused Silica Capillary Tubing, Non-Deactivated, 363 um OD, 75 um ID, Custom