Trajan SGE 062454 Fused Silica Capillary Tubing, Non-Deactivated, 170 um OD, 110 um ID, 10 m L; 1/Ea

Representative image only.

Trajan SGE 062454 Fused Silica Capillary Tubing, Non-Deactivated, 170 um OD, 110 um ID, 10 m L; 1/Ea

Trajan | Mfr: 062454| Item: 1600662

Precision Tubing for Performance

₹15,400.00INR / Each

₹15,400.00INR / Each
Product Description

Key Features

  • High homogeneity
  • Chemical inertness
  • High intrinsic tensile strength
  • Pressure resistant to 1000 bar
  • 6 month warranty

More About this Item

Coated fused silica tubing exhibits excellent flexibility, chemical stability, tensile strength and exacting tolerances making it the tubing of choice for GC capillary applications, LC tubing and other micro tubing requirements.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

6 Month Warranty
Specifications
  • DescriptionTrajan SGE Fused Silica Capillary Tubing, Non-Deactivated, 170 um OD, 110 um ID, 10 m L; 1/Ea
  • Warranty6 month
Reviews
Q&A