Trajan SGE 062447 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 220 um ID, 5 m L; 1/Ea
Representative image only.
Trajan SGE 062447 Fused Silica Capillary Tubing, Methyl Deactivated, 363 um OD, 220 um ID, 5 m L; 1/Ea
Trajan | Mfr: 062447| Item: 1600650
Precision Tubing for Performance
₹11,200.00INR / Each
₹11,200.00INR / Each
Product Description
Key Features
  • High homogeneity
  • Chemical inertness
  • High intrinsic tensile strength
  • Pressure resistant to 1000 bar
  • 6 month warranty
More About this Item
Coated fused silica tubing exhibits excellent flexibility, chemical stability, tensile strength and exacting tolerances making it the tubing of choice for GC capillary applications, LC tubing and other micro tubing requirements.