Trajan SGE 0624250 PEEKsil Tubing, 1/16" OD, 0.05 mm ID, 500 mm Length; 2/Pk
Representative image only.
Trajan SGE 0624250 PEEKsil Tubing, 1/16" OD, 0.05 mm ID, 500 mm Length; 2/Pk
Trajan | Mfr: 0624250| Item: 1600582
Fused silica lined PEEK
₹10,800.00INR / Pkg of 2
₹10,800.00INR / Pkg of 2
Product Description
Key Features
  • Inert smooth flow path to minimize band broadening
  • Precise inner bore diameter
  • Flexible and robust
  • Color coded for easy identification of internal diameter
  • 6 month warranty
More About this Item
Compatible with most organic solvents, reduces dispersion and loss of efficiency, sensitivty, and resolution
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE PEEKsil Tubing, 1/16" OD, 0.05 mm ID, 500 mm Length; 2/Pk