Trajan SGE 0418992 High Temperature Silicone Septa, 12.5 mm Diameter; 24/Pk
Representative image only.
Trajan SGE 0418992 High Temperature Silicone Septa, 12.5 mm Diameter; 24/Pk
Trajan | Mfr: 0418992| Item: 1600525
Optimized for the most demanding GC and GC/MS applications
₹8,600.00INR / Pkg of 24
₹8,600.00INR / Pkg of 24
Product Description
Key Features
  • Formulated to extend low-bleed and outstanding mechanical properties
  • 6 month warranty
More About this Item
(Bleed and Temperature Optimized) are designed for low bleed and temperatures up to 400°C (752 °F). Septa changes are easy—septa will not stick to your port.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE High Temperature Silicone Septa, 12.5 mm Diameter; 24/Pk