Trajan SGE 041895 High Temperature Silicone Septa, Compatible with Shimadzu, Plug Style; 50/Pk
Representative image only.
Trajan SGE 041895 High Temperature Silicone Septa, Compatible with Shimadzu, Plug Style; 50/Pk
Trajan | Mfr: 298735| Item: 1600519
Optimized for the most demanding GC and GC/MS applications
₹17,100.00INR / Pkg of 50
₹17,100.00INR / Pkg of 50
Product Description
Key Features
  • Formulated to extend low-bleed and outstanding mechanical properties
  • 6 month warranty
More About this Item
(Bleed and Temperature Optimized) are designed for low bleed and temperatures up to 400°C (752 °F). Septa changes are easy—septa will not stick to your port.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE High Temperature Silicone Septa, Compatible with Shimadzu, Plug Style; 50/Pk