Trajan SGE 041890 Enduro Septa, Compatible with Shimadzu, Plug Style, Blue Color; 50/Pk
Representative image only.
Trajan SGE 041890 Enduro Septa, Compatible with Shimadzu, Plug Style, Blue Color; 50/Pk
Trajan | Mfr: 234121| Item: 1600518
Give your GC the seal of approval
₹18,200.00INR / Pkg of 50
₹18,200.00INR / Pkg of 50
Product Description
Key Features
  • Reliably seal against carrier gas pressure
  • Capable of being pierced and resealing time after time without septa deposition in the GC inlet
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Enduro Septa, Compatible with Shimadzu, Plug Style, Blue Color; 50/Pk