Trajan SGE 041881 High Temperature Silicone Septa, 9.5 mm Diameter; 100/Pk
Representative image only.
Trajan SGE 041881 High Temperature Silicone Septa, 9.5 mm Diameter; 100/Pk
Trajan | Mfr: 298705| Item: 1600516
Optimized for the most demanding GC and GC/MS applications
₹28,800.00INR / Pkg of 100
₹28,800.00INR / Pkg of 100
Product Description
Key Features
  • Formulated to extend low-bleed and outstanding mechanical properties
  • 6 month warranty
More About this Item
(Bleed and Temperature Optimized) are designed for low bleed and temperatures up to 400°C (752 °F). Septa changes are easy—septa will not stick to your port.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE High Temperature Silicone Septa, 9.5 mm Diameter; 100/Pk