Trajan SGE 041824 General Purpose Septa, 9 mm Diameter; 50/Pk
Representative image only.
Trajan SGE 041824 General Purpose Septa, 9 mm Diameter; 50/Pk
Trajan | Mfr: 234108| Item: 1600500
Give your GC the seal of approval
₹6,500.00INR / Pkg of 50
₹6,500.00INR / Pkg of 50
Product Description
Key Features
  • Reliably seal against carrier gas pressure
  • Capable of being pierced and resealing time after time without septa deposition in the GC inlet
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE General Purpose Septa, 9 mm Diameter; 50/Pk