Trajan SGE 039807 Replacement Luer Lock Needle, 70 mm, 0.63 mm OD, 23 G, Bevel Tip; 5/Pk
Representative image only.
Trajan SGE 039807 Replacement Luer Lock Needle, 70 mm, 0.63 mm OD, 23 G, Bevel Tip; 5/Pk
Trajan | Mfr: 039807| Item: 1600480
Wide range of needles available to suit t SGE 5 µL, 10 µL, 25-500 µL, 1-2.5 mL, 5-10 mL, Luer Lock and NanoVolume syringes
₹8,000.00INR / Pkg of 5
₹8,000.00INR / Pkg of 5
Product Description
Key Features
  • Stainless Steel
  • Available in various lengths and OD configurations
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Replacement Luer Lock Needle, 70 mm, 0.63 mm OD, 23 G, Bevel Tip; 5/Pk