Trajan SGE 008975 Headspace/Soil Gas Syringes, 10 mL, 1/Ea
Representative image only.
Trajan SGE 008975 Headspace/Soil Gas Syringes, 10 mL, 1/Ea
Trajan | Mfr: 008975| Item: 1600194
Ideal for soil headspace extraction and sample storage prior to injection
₹25,600.00INR / Each
₹25,600.00INR / Each
Product Description
Key Features
  • Ready to use gas tight syringe
  • Efficient sample intake
  • PTFE plunger tip for perfect sealing
  • Store gas in the syringe without contamination
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Headspace/Soil Gas Syringes, 10 mL, 1/Ea