Trajan SGE 008160 Gas Tight Syringe with Pre-fitted Luer Lock Valve, 1 mL; 1/Ea
Representative image only.
Trajan SGE 008160 Gas Tight Syringe with Pre-fitted Luer Lock Valve, 1 mL; 1/Ea
Trajan | Mfr: 008160| Item: 1600166
The comprehensive range of SGE syringes are designed and tested to meet critical specifications. We combine precision manufacturing of components and meticulous assembly, to produce syringes of the highest quality.
₹15,500.00INR / Each
₹15,500.00INR / Each
Product Description
Key Features
  • Minimal carryover
  • Gas tight
  • PTFE Plunger tip
  • Resistant to a wide range of solvents
  • 6 month warranty
More About this Item
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE Gas Tight Syringe with Pre-fitted Luer Lock Valve, 1 mL; 1/Ea