Trajan SGE 005292 MEPS Syringe, Compatible with Agilent 7693A, Removable Needle, 100 uL, 54.1 mm Scale; 1/Ea
Representative image only.
Trajan SGE 005292 MEPS Syringe, Compatible with Agilent 7693A, Removable Needle, 100 uL, 54.1 mm Scale; 1/Ea
Trajan | Mfr: 005292| Item: 1600084
Extraction to injection in a single process
₹19,700.00INR / Each
₹19,700.00INR / Each
Product Description
Key Features
  • Compatible for use with MEPS BINs
  • Save hours in sample preparation
  • Reduce solvent usage and sample volume
  • 6 month warranty
More About this Item
MEPS Barrel Insert and Needle (BIN) assemble contains the stationary phase, and is built into the syringe needle. Use requires both a suitable syringe and a MEPS BIN
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
6 Month Warranty
Specifications
  • DescriptionTrajan SGE MEPS Syringe, Compatible with Agilent 7693A, Removable Needle, 100 uL, 54.1 mm Scale; 1/Ea